missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Phenyl p-tolyl sulfone, 98+%, Thermo Scientific Chemicals
$139.79 - $416.03
Chemical Identifiers
CAS | 640-57-3 |
---|---|
Molecular Formula | C13H12O2S |
Molecular Weight (g/mol) | 232.297 |
MDL Number | MFCD00025038 |
InChI Key | YBRXHRWLEFCFEG-UHFFFAOYSA-N |
Synonym | phenyl p-tolyl sulfone, sulfone, phenyl p-tolyl, 4-methylphenyl phenyl sulfone, benzene, 1-methyl-4-phenylsulfonyl, 1-methyl-4-phenylsulfonyl benzene, 4-methyldiphenylsulfone, p-tolyl phenyl sulfone, p-phenylsulphonyl toluene, 1-benzenesulfonyl-4-methylbenzene, 1-benzenesulfonyl-4-methyl-benzene |
PubChem CID | 69497 |
IUPAC Name | 1-(benzenesulfonyl)-4-methylbenzene |
SMILES | CC1=CC=C(C=C1)S(=O)(=O)C2=CC=CC=C2 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0312809
|
Thermo Scientific Chemicals
L0312809 |
10 g |
Each for $139.79
|
|
|||||
AAL0312818
|
Thermo Scientific Chemicals
L0312818 |
50 g |
Each for $416.03
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
640-57-3 | |
232.297 | |
YBRXHRWLEFCFEG-UHFFFAOYSA-N | |
69497 | |
CC1=CC=C(C=C1)S(=O)(=O)C2=CC=CC=C2 |
C13H12O2S | |
MFCD00025038 | |
phenyl p-tolyl sulfone, sulfone, phenyl p-tolyl, 4-methylphenyl phenyl sulfone, benzene, 1-methyl-4-phenylsulfonyl, 1-methyl-4-phenylsulfonyl benzene, 4-methyldiphenylsulfone, p-tolyl phenyl sulfone, p-phenylsulphonyl toluene, 1-benzenesulfonyl-4-methylbenzene, 1-benzenesulfonyl-4-methyl-benzene | |
1-(benzenesulfonyl)-4-methylbenzene |
Specifications
640-57-3 | |
Odorless | |
MFCD00025038 | |
1872872 | |
YBRXHRWLEFCFEG-UHFFFAOYSA-N | |
1-(benzenesulfonyl)-4-methylbenzene | |
69497 | |
≥98% |
125°C to 129°C | |
C13H12O2S | |
10 g | |
phenyl p-tolyl sulfone, sulfone, phenyl p-tolyl, 4-methylphenyl phenyl sulfone, benzene, 1-methyl-4-phenylsulfonyl, 1-methyl-4-phenylsulfonyl benzene, 4-methyldiphenylsulfone, p-tolyl phenyl sulfone, p-phenylsulphonyl toluene, 1-benzenesulfonyl-4-methylbenzene, 1-benzenesulfonyl-4-methyl-benzene | |
CC1=CC=C(C=C1)S(=O)(=O)C2=CC=CC=C2 | |
232.297 | |
232.3 | |
Phenyl p-tolyl sulfone |
Safety and Handling
EINECSNumber : 211-364-7
RTECSNumber : WR6880000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only