missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phenyl phosphate, disodium salt dihydrate, 98%, Thermo Scientific Chemicals

Phenyl phosphate, disodium salt dihydrate, 98%, C6H5Na2O4P.2H2O, CAS Number-66778-08-3, sodiumphenylphosphatedibasicdihydrate, 10g, White, 221-917-4, 254.09, HYGROSCOPIC, UMRLIPYPVZYYFT-UHFFFAOYSA-N, Authentic, phenyl dihydrogen phosphate;sodium;dihydrate, C6H5OP(O)(ONa)2.2H2O, >300.0 deg.C

Supplier:  Thermo Scientific Chemicals 211830100

 View more versions of this product

Catalog No. AC211830100


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Phenyl phosphate, disodium salt dihydrate
66778-08-3
White
98%
C6H5Na2O42H2O
MFCD00149080
14, 3357
Solubility in water: soluble. Other solubilities: soluble in ethanol and chloroform,insoluble in acetone and ether
C1=CC=C(C=C1)OP(=O)(O)O.O.O.[Na].[Na]
254.09
254.09
Solid
98%
>300.0°C
Authentic
Glass bottle
C6H5OP(O)(ONa)2·2H2O
10 g
sodiumphenylphosphatedibasicdihydrate
UMRLIPYPVZYYFT-UHFFFAOYSA-N
phenyl dihydrogen phosphate;sodium;dihydrate
131883070
98%

Chemical Identifiers

66778-08-3
254.09
UMRLIPYPVZYYFT-UHFFFAOYSA-N
131883070
C1=CC=C(C=C1)OP(=O)(O)O.O.O.[Na].[Na]
C6H5Na2O42H2O
MFCD00149080
sodiumphenylphosphatedibasicdihydrate
phenyl dihydrogen phosphate;sodium;dihydrate
Videos
Safety and Handling

Safety and Handling

HYGROSCOPIC

EINECSNumber : 221-917-4

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only