missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phenyl phosphate, disodium salt dihydrate, 98%, Thermo Scientific Chemicals

Phenyl phosphate, disodium salt dihydrate, 98%, C6H5Na2O4P.2H2O, CAS Number-66778-08-3, sodiumphenylphosphatedibasicdihydrate, 10g, White, 221-917-4, 254.09, HYGROSCOPIC, UMRLIPYPVZYYFT-UHFFFAOYSA-N, Authentic, phenyl dihydrogen phosphate;sodium;dihydrate, C6H5OP(O)(ONa)2.2H2O, >300.0 deg.C

$95.44 - $408.89

Chemical Identifiers

CAS 66778-08-3
Molecular Formula C6H5Na2O42H2O
Molecular Weight (g/mol) 254.09
MDL Number MFCD00149080
InChI Key UMRLIPYPVZYYFT-UHFFFAOYSA-N
Synonym sodiumphenylphosphatedibasicdihydrate
PubChem CID 131883070
IUPAC Name phenyl dihydrogen phosphate;sodium;dihydrate
SMILES C1=CC=C(C=C1)OP(=O)(O)O.O.O.[Na].[Na]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC211830100
SDS View Documents
Thermo Scientific Chemicals
211830100
10 g Glass bottle
Each for $95.44
 
AC211831000
SDS View Documents
Thermo Scientific Chemicals
211831000
100 g Glass bottle
Each for $408.89
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

66778-08-3
254.09
UMRLIPYPVZYYFT-UHFFFAOYSA-N
131883070
C1=CC=C(C=C1)OP(=O)(O)O.O.O.[Na].[Na]
C6H5Na2O42H2O
MFCD00149080
sodiumphenylphosphatedibasicdihydrate
phenyl dihydrogen phosphate;sodium;dihydrate

Specifications

66778-08-3
White
98%
C6H5Na2O42H2O
MFCD00149080
14, 3357
Solubility in water: soluble. Other solubilities: soluble in ethanol and chloroform,insoluble in acetone and ether
C1=CC=C(C=C1)OP(=O)(O)O.O.O.[Na].[Na]
254.09
254.09
Solid
>300.0°C
Authentic
Glass bottle
C6H5OP(O)(ONa)2·2H2O
10 g
sodiumphenylphosphatedibasicdihydrate
UMRLIPYPVZYYFT-UHFFFAOYSA-N
phenyl dihydrogen phosphate;sodium;dihydrate
131883070
98%
Phenyl phosphate, disodium salt dihydrate, 98%
Videos
Safety and Handling

Safety and Handling

HYGROSCOPIC

EINECSNumber : 221-917-4

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only