Learn More
Phenyl phosphate, disodium salt dihydrate, 98%, Thermo Scientific Chemicals
Phenyl phosphate, disodium salt dihydrate, 98%, C6H5Na2O4P.2H2O, CAS Number-66778-08-3, sodiumphenylphosphatedibasicdihydrate, 10g, White, 221-917-4, 254.09, HYGROSCOPIC, UMRLIPYPVZYYFT-UHFFFAOYSA-N, Authentic, phenyl dihydrogen phosphate;sodium;dihydrate, C6H5OP(O)(ONa)2.2H2O, >300.0 deg.C
Supplier: Thermo Scientific Chemicals 211831000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Phenyl phosphate, disodium salt dihydrate | |
66778-08-3 | |
White | |
98% | |
C6H5Na2O4P·2H2O | |
MFCD00149080 | |
14, 3357 | |
Solubility in water: soluble. Other solubilities: soluble in ethanol and chloroform,insoluble in acetone and ether | |
C1=CC=C(C=C1)OP(=O)(O)O.O.O.[Na].[Na] | |
254.09 | |
254.09 | |
Crystalline Powder |
98% | |
>300.0°C | |
Authentic | |
Glass bottle | |
C6H5OP(O)(ONa)2·2H2O | |
100 g | |
sodiumphenylphosphatedibasicdihydrate | |
UMRLIPYPVZYYFT-UHFFFAOYSA-N | |
phenyl dihydrogen phosphate;sodium;dihydrate | |
131883070 | |
98% |
Chemical Identifiers
66778-08-3 | |
254.09 | |
UMRLIPYPVZYYFT-UHFFFAOYSA-N | |
131883070 | |
C1=CC=C(C=C1)OP(=O)(O)O.O.O.[Na].[Na] |
C6H5Na2O4P·2H2O | |
MFCD00149080 | |
sodiumphenylphosphatedibasicdihydrate | |
phenyl dihydrogen phosphate;sodium;dihydrate |
Safety and Handling
HYGROSCOPIC
EINECSNumber : 221-917-4
TSCA : TSCA
RUO – Research Use Only