missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phenylacetyl chloride, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 130301000

 View more versions of this product

Catalog No. AC130301000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Phenylacetyl chloride
103-80-0
94.0°C to 95.0°C (12.0 mmHg)
97.5% min. (GC)
C8H7ClO
C6H5CH2COCl
100 g
1.16
Solubility in water: reacts with water
ClC(=O)CC1=CC=CC=C1
154.59
154.6
98%
1.1600g/mL
102°C
Glass bottle
1.5320 to 1.5330
MFCD00000729
09,436
phenylacetyl chloride, benzeneacetyl chloride, acetyl chloride, phenyl, phenylacetic acid chloride, phenacetyl chloride, phenylacetylchloride, unii-t30899drnd, phenyl acetyl chloride, a-toluoyl chloride, phenylacetyl
VMZCDNSFRSVYKQ-UHFFFAOYSA-N
2-phenylacetyl chloride
7679
98%

Chemical Identifiers

103-80-0
154.59
VMZCDNSFRSVYKQ-UHFFFAOYSA-N
7679
ClC(=O)CC1=CC=CC=C1
C8H7ClO
MFCD00000729
phenylacetyl chloride, benzeneacetyl chloride, acetyl chloride, phenyl, phenylacetic acid chloride, phenacetyl chloride, phenylacetylchloride, unii-t30899drnd, phenyl acetyl chloride, a-toluoyl chloride, phenylacetyl
2-phenylacetyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 203-146-5

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only