missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phenylacetylene, 98%, pure, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 152460250

 View more versions of this product

Catalog No. AC152460250



Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Phenylacetylene
-44.8°C
0.9300g/mL
31°C
97.5% min. (GC)
C8H6
C6H5C≡CH
05,511
15,391
Solubility in water: insoluble. Other solubilities: miscible with alcohol,ether and other organic,solvents
C#CC1=CC=CC=C1
102.14
102.14
Pure
536-74-3
Colorless to Yellow
142.0°C to 144.0°C
Authentic
Glass bottle
1.5470 to 1.5500
25 g
0.93
phenylacetylene, phenylethyne, benzene, ethynyl, 1-phenylethyne, phenylacetylide, phenyl acetylene, ethyne, phenyl, acetylene, phenyl, ethynyl-benzene, phenylacethylene
UEXCJVNBTNXOEH-UHFFFAOYSA-N
ethynylbenzene
10821
98%
Liquid

Chemical Identifiers

536-74-3
102.14
phenylacetylene, phenylethyne, benzene, ethynyl, 1-phenylethyne, phenylacetylide, phenyl acetylene, ethyne, phenyl, acetylene, phenyl, ethynyl-benzene, phenylacethylene
ethynylbenzene
C8H6
UEXCJVNBTNXOEH-UHFFFAOYSA-N
10821
C#CC1=CC=CC=C1
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapor.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.

GHS Signal Word: Danger

EINECSNumber : 208-645-1

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only