missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phenylacetylene, 98%, pure, Thermo Scientific Chemicals

$98.83 - $227.53

Chemical Identifiers

CAS 536-74-3
Molecular Formula C8H6
Molecular Weight (g/mol) 102.14
InChI Key UEXCJVNBTNXOEH-UHFFFAOYSA-N
Synonym phenylacetylene, phenylethyne, benzene, ethynyl, 1-phenylethyne, phenylacetylide, phenyl acetylene, ethyne, phenyl, acetylene, phenyl, ethynyl-benzene, phenylacethylene
PubChem CID 10821
IUPAC Name ethynylbenzene
SMILES C#CC1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC152460250
SDS View Documents
Thermo Scientific Chemicals
152460250
25 g Glass bottle
Each for $98.83
Case of 6 Each for $0.00
 
AC152461000
SDS View Documents
Thermo Scientific Chemicals
152461000
100 g Glass bottle
Each for $227.53
Case of 6 Each for $0.00
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

536-74-3
102.14
phenylacetylene, phenylethyne, benzene, ethynyl, 1-phenylethyne, phenylacetylide, phenyl acetylene, ethyne, phenyl, acetylene, phenyl, ethynyl-benzene, phenylacethylene
ethynylbenzene
C8H6
UEXCJVNBTNXOEH-UHFFFAOYSA-N
10821
C#CC1=CC=CC=C1

Specifications

536-74-3
Colorless to Yellow
142.0°C to 144.0°C
Authentic
Glass bottle
1.5470 to 1.5500
25 g
0.93
phenylacetylene, phenylethyne, benzene, ethynyl, 1-phenylethyne, phenylacetylide, phenyl acetylene, ethyne, phenyl, acetylene, phenyl, ethynyl-benzene, phenylacethylene
UEXCJVNBTNXOEH-UHFFFAOYSA-N
ethynylbenzene
10821
98%
Liquid
-44.8°C
0.9300g/mL
31°C
97.5% min. (GC)
C8H6
C6H5C≡CH
05,511
15,391
Solubility in water: insoluble. Other solubilities: miscible with alcohol,ether and other organic,solvents
C#CC1=CC=CC=C1
102.14
102.14
Pure
Phenylacetylene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapor.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.

GHS Signal Word: Danger

EINECSNumber : 208-645-1

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only