Phenylalanine, USP, 98.5-101.5%, Spectrum™ Chemical
P1582, 63-91-2, C9H11NO2
Supplier:  Spectrum Chemical Mfg Cor P1582100GM
 View more versions of this product
Catalog No. 18605122
Explore available promotions
Description
Description
Spectrum™ Chemical Phenylalanine, USP is an amino acid, a "building block" of protein. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
63-91-2
5.4 to 6.0
0.004
3e-005
Poly Bottle
NC(CC1=CC=CC=C1)C(O)=O
-32.7° to -34.7°
0.0003
USP
1
0.0005
C9H11NO2
0.003
COLNVLDHVKWLRT-UHFFFAOYNA-N
2-amino-3-phenylpropanoic acid
165.19
98.5 to 101.5%
100 g
Chemical Identifiers
63-91-2
165.19
2-amino-3-phenylpropanoic acid
C9H11NO2
COLNVLDHVKWLRT-UHFFFAOYNA-N
NC(CC1=CC=CC=C1)C(O)=O