missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Phenylalanine, USP, 98.5-101.5%, Spectrum™ Chemical
P1582, 63-91-2, C9H11NO2
$167.73 - $3,533.27
Chemical Identifiers
CAS | 63-91-2 |
---|---|
Molecular Formula | C9H11NO2 |
Molecular Weight (g/mol) | 165.19 |
InChI Key | COLNVLDHVKWLRT-UHFFFAOYNA-N |
IUPAC Name | 2-amino-3-phenylpropanoic acid |
SMILES | NC(CC1=CC=CC=C1)C(O)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18605121
|
Spectrum Chemical Mfg Cor
P158225GM |
25 g | Poly Bottle |
N/A
|
|
||||
18605122
|
Spectrum Chemical Mfg Cor
P1582100GM |
100 g | Poly Bottle |
N/A
|
|
||||
18605123
|
Spectrum Chemical Mfg Cor
P15821KG |
1 kg | Poly Pail |
N/A
|
|
||||
18605124
|
Spectrum Chemical Mfg Cor
P15825KGBL |
5 kg | Poly Pail |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
18605125
|
Spectrum Chemical Mfg Cor
P158250KGBL |
50 kg | Fiber Drum |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Phenylalanine, USP is an amino acid, a "building block" of protein. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
63-91-2 | |
165.19 | |
2-amino-3-phenylpropanoic acid |
C9H11NO2 | |
COLNVLDHVKWLRT-UHFFFAOYNA-N | |
NC(CC1=CC=CC=C1)C(O)=O |
Specifications
63-91-2 | |
5.4 to 6.0 | |
0.004 | |
3e-005 | |
Poly Bottle | |
NC(CC1=CC=CC=C1)C(O)=O | |
-32.7° to -34.7° | |
0.0003 | |
USP |
1 | |
0.0005 | |
C9H11NO2 | |
0.003 | |
COLNVLDHVKWLRT-UHFFFAOYNA-N | |
2-amino-3-phenylpropanoic acid | |
165.19 | |
98.5 to 101.5% | |
25 g |