missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phenylmagnesium bromide, 1.6M solution in CPME, AcroSeal™, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 445961000

 View more versions of this product

Catalog No. AC445961000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Phenylmagnesium bromide
−1°C
C6H5BrMg
MFCD00000038
phenylmagnesium bromide, grignard reagent, magnesium, bromophenyl, magnesium, bromophenyl, phenylmagnesiumbromide, bromo phenyl magnesium, bromo phenyl magnesium, phenylmagnesium bromide solution, 3.0 m in diethyl ether, bromomagnesiobenzene, phenyhnagnesium bromide, phenyhnagnesium bromide
ANRQGKOBLBYXFM-UHFFFAOYSA-M
magnesium;benzene;bromide
66852
100-58-3,5614-37-9
AcroSeal™ Glass bottle
C6H5MgBr
100 mL
Solubility in water: reacts
Br[Mg]C1=CC=CC=C1
181.32
181.31

Chemical Identifiers

100-58-3
181.32
ANRQGKOBLBYXFM-UHFFFAOYSA-M
66852
Br[Mg]C1=CC=CC=C1
C6H5BrMg
MFCD00000038
phenylmagnesium bromide, grignard reagent, magnesium, bromophenyl, magnesium, bromophenyl, phenylmagnesiumbromide, bromo phenyl magnesium, bromo phenyl magnesium, phenylmagnesium bromide solution, 3.0 m in diethyl ether, bromomagnesiobenzene, phenyhnagnesium bromide, phenyhnagnesium bromide
magnesium;benzene;bromide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapor.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful to aquatic life with long lasting effects.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

GHS Signal Word: Danger

EINECSNumber : 202-867-2

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only