missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phenyltrimethylammonium chloride, 98+%, Thermo Scientific Chemicals

$86.92 - $294.01

Chemical Identifiers

CAS 138-24-9
Molecular Formula C9H14ClN
Molecular Weight (g/mol) 171.67
MDL Number MFCD00011790
InChI Key MQAYPFVXSPHGJM-UHFFFAOYSA-M
Synonym phenyltrimethylammonium chloride, n,n,n-trimethylbenzenaminium chloride, trimethylphenylammonium chloride, n,n,n-trimethylanilinium chloride, ammonyx 200, benzenaminium, n,n,n-trimethyl-, chloride, trimethylphenylammoniumchloride, trimethylanilinium chloride, ammonium, phenyltrimethyl-, chloride, trimethylphenyl ammonium chloride
PubChem CID 67309
SMILES [Cl-].C[N+](C)(C)C1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2285122
View Documents
Thermo Scientific Chemicals
B2285122
100 g
Each for $86.92
 
AAB2285136
View Documents
Thermo Scientific Chemicals
B2285136
500 g
Each for $294.01
 
Description

Description

Phenyltrimethylammonium chloride is an effective phase transfer catalyst, methylating agent, also useful for alkaline depolymerizations, corrosion inhibitor for low carbon steel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenyltrimethylammonium chloride is an effective phase transfer catalyst, methylating agent, also useful for alkaline depolymerizations, corrosion inhibitor for low carbon steel.

Solubility
Soluble in water 333 g/L.

Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agent, water/ moisture.
Specifications

Chemical Identifiers

138-24-9
171.67
MQAYPFVXSPHGJM-UHFFFAOYSA-M
67309
C9H14ClN
MFCD00011790
phenyltrimethylammonium chloride, n,n,n-trimethylbenzenaminium chloride, trimethylphenylammonium chloride, n,n,n-trimethylanilinium chloride, ammonyx 200, benzenaminium, n,n,n-trimethyl-, chloride, trimethylphenylammoniumchloride, trimethylanilinium chloride, ammonium, phenyltrimethyl-, chloride, trimethylphenyl ammonium chloride
[Cl-].C[N+](C)(C)C1=CC=CC=C1

Specifications

138-24-9
C9H14ClN
100 g
3572527
phenyltrimethylammonium chloride, n,n,n-trimethylbenzenaminium chloride, trimethylphenylammonium chloride, n,n,n-trimethylanilinium chloride, ammonyx 200, benzenaminium, n,n,n-trimethyl-, chloride, trimethylphenylammoniumchloride, trimethylanilinium chloride, ammonium, phenyltrimethyl-, chloride, trimethylphenyl ammonium chloride
MQAYPFVXSPHGJM-UHFFFAOYSA-M
N,N,N-trimethylanilinium chloride
67309
≥98%
∼236°C (sublimation)
MFCD00011790
UN2811
Hygroscopic
Soluble in water 333g/L.
[Cl-].C[N+](C)(C)C1=CC=CC=C1
171.67
171.67
Phenyltrimethylammonium chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311
Toxic if swallowed.
Toxic in contact with skin.

P264b-P270-P280-P301+P310-P302+P352-P312-P330-P361-P363-P501c

H301+H311

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 205-319-0

RTECSNumber : BT2190000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only