missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phosphoenolpyruvic acid mono(cyclohexylammonium) salt, 98%, Thermo Scientific Chemicals

$153.15 - $2243.60

Chemical Identifiers

CAS 10526-80-4
Molecular Formula C9H18NO6P
Molecular Weight (g/mol) 267.22
MDL Number MFCD00036375
InChI Key VHFCNZDHPABZJO-UHFFFAOYSA-N
Synonym cyclohexanamine 2-phosphonooxy acrylate, phosphoenolpyruvic acid cyclohexylammonium salt, 2-propenoic acid, 2-phosphonooxy-, compd. with cyclohexanamine 1:1, cyclohexylamine; phosphoenolpyruvic acid, phosphoenolpyruvic acid cyclohexylamine salt, phosphoenolpyruvic acid, cyclohexylammonium salt, cyclohexanamine; 2-phosphonooxyprop-2-enoic acid, phospho enol pyruvic acid cyclohexylammonium salt, phosphoenolpyruvic acid monocyclohexylammonium salt
PubChem CID 82702
IUPAC Name cyclohexanamine;2-phosphonooxyprop-2-enoic acid
SMILES NC1CCCCC1.OC(=O)C(=C)OP(O)(O)=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1238503
View Documents
Thermo Scientific Chemicals
A1238503
1 g
Each for $153.15
 
AAA1238506
View Documents
Thermo Scientific Chemicals
A1238506
5 g
Each for $543.67
 
AAA1238514
View Documents
Thermo Scientific Chemicals
A1238514
25 g
Each for $2,243.60
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10526-80-4
267.22
VHFCNZDHPABZJO-UHFFFAOYSA-N
82702
NC1CCCCC1.OC(=O)C(=C)OP(O)(O)=O
C9H18NO6P
MFCD00036375
cyclohexanamine 2-phosphonooxy acrylate, phosphoenolpyruvic acid cyclohexylammonium salt, 2-propenoic acid, 2-phosphonooxy-, compd. with cyclohexanamine 1:1, cyclohexylamine; phosphoenolpyruvic acid, phosphoenolpyruvic acid cyclohexylamine salt, phosphoenolpyruvic acid, cyclohexylammonium salt, cyclohexanamine; 2-phosphonooxyprop-2-enoic acid, phospho enol pyruvic acid cyclohexylammonium salt, phosphoenolpyruvic acid monocyclohexylammonium salt
cyclohexanamine;2-phosphonooxyprop-2-enoic acid

Specifications

10526-80-4
C9H18NO6P
1 g
VHFCNZDHPABZJO-UHFFFAOYSA-N
cyclohexanamine;2-phosphonooxyprop-2-enoic acid
82702
98%
∼140°C (decomposition)
MFCD00036375
cyclohexanamine 2-phosphonooxy acrylate, phosphoenolpyruvic acid cyclohexylammonium salt, 2-propenoic acid, 2-phosphonooxy-, compd. with cyclohexanamine 1:1, cyclohexylamine; phosphoenolpyruvic acid, phosphoenolpyruvic acid cyclohexylamine salt, phosphoenolpyruvic acid, cyclohexylammonium salt, cyclohexanamine; 2-phosphonooxyprop-2-enoic acid, phospho enol pyruvic acid cyclohexylammonium salt, phosphoenolpyruvic acid monocyclohexylammonium salt
NC1CCCCC1.OC(=O)C(=C)OP(O)(O)=O
267.22
267.22
Phosphoenolpyruvic acid mono (cyclohexylammonium) salt
Safety and Handling

Safety and Handling

EINECSNumber : 234-084-7

TSCA : Yes

Recommended Storage : Store at -20°C

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only