Learn More
Phosphorus pentoxide, 99+%, for analysis, Thermo Scientific Chemicals
$75.48 - $225.63
Chemical Identifiers
CAS | 1314-56-3 |
---|---|
Molecular Formula | O5P2 |
Molecular Weight (g/mol) | 141.94 |
MDL Number | MFCD00011440 |
InChI Key | DLYUQMMRRRQYAE-UHFFFAOYSA-N |
Synonym | Phosphoric anhydride |
IUPAC Name | tricyclo[3.3.1.1³,â·]tetraphosphoxane-1,3,5,7-tetrone |
SMILES | O=P12OP3(=O)OP(=O)(O1)OP(=O)(O2)O3 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC215751000
|
Thermo Scientific Chemicals
215751000 |
100 g | Glass bottle |
N/A
|
|
|||||
AC215755000
|
Thermo Scientific Chemicals
215755000 |
500 g | Glass bottle |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
AdsorbentChemical Identifiers
1314-56-3 | |
141.94 | |
DLYUQMMRRRQYAE-UHFFFAOYSA-N | |
tricyclo[3.3.1.1³,â·]tetraphosphoxane-1,3,5,7-tetrone |
O5P2 | |
MFCD00011440 | |
Phosphoric anhydride | |
O=P12OP3(=O)OP(=O)(O1)OP(=O)(O2)O3 |
Specifications
340.0°C to 360.0°C | |
White | |
100 g | |
O5P2 | |
MFCD00011440 | |
15, 7467 | |
Solubility in water: 85% (20°C); reacts forming h3po4 with evol.of heat. | |
O=P12OP3(=O)OP(=O)(O1)OP(=O)(O2)O3 | |
141.94 | |
≥99% | |
Glass bottle | |
Phosphorus pentoxide, p.a. |
1314-56-3 | |
Very Deliquescing Powder | |
99+% | |
P2O5 | |
01,871; 07,291; 10,319; 14,151; 15,267 | |
Phosphoric anhydride | |
DLYUQMMRRRQYAE-UHFFFAOYSA-N | |
tricyclo[3.3.1.1³,â·]tetraphosphoxane-1,3,5,7-tetrone | |
141.94 | |
Analytical | |
2.39 |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Reacts violently with water.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 215-236-1
RTECSNumber : TH3945000
TSCA : TSCA
RUO – Research Use Only