missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phosphorus tribromide, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 169471000

 View more versions of this product

Catalog No. AC169471000

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
Specifications

Specifications

175.0°C
-40.0°C
Brown or Colorless to Yellow
100 g
Br3P
MFCD00011436
15, 7469
Solubility in water: reacts. Other solubilities: soluble in acetone,alcohol,carbon disulfide,,hydrogen sulfide,methanol,ether,chloroform and,carbon tetrachloride
P(Br)(Br)Br
270.69
270.69
Glass bottle
2.88
Phosphorus tribromide
7789-60-8
Liquid
98.5% min. (ex Br) (Argentometry)
PBr3
01,873; 02,330; 07,292
phosphorus tribromide, phosphorous tribromide, tribromophosphine, phosphorus iii bromide, phosphorous bromide, pbr3, phosphorus bromide, phosphorus bromide pbr3, unii-58r3866pua, tribro-mophosphine
IPNPIHIZVLFAFP-UHFFFAOYSA-N
tribromophosphane
24614
99%
1.697
2.8800g/mL

Chemical Identifiers

7789-60-8
270.69
IPNPIHIZVLFAFP-UHFFFAOYSA-N
24614
P(Br)(Br)Br
Br3P
MFCD00011436
phosphorus tribromide, phosphorous tribromide, tribromophosphine, phosphorus iii bromide, phosphorous bromide, pbr3, phosphorus bromide, phosphorus bromide pbr3, unii-58r3866pua, tribro-mophosphine
tribromophosphane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Reacts violently with water.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 232-178-2

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only