missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phosphorus tribromide, 99%, Thermo Scientific Chemicals

$116.26 - $293.74

Chemical Identifiers

CAS 7789-60-8
Molecular Formula Br3P
Molecular Weight (g/mol) 270.69
MDL Number MFCD00011436
InChI Key IPNPIHIZVLFAFP-UHFFFAOYSA-N
Synonym phosphorus tribromide, phosphorous tribromide, tribromophosphine, phosphorus iii bromide, phosphorous bromide, pbr3, phosphorus bromide, phosphorus bromide pbr3, unii-58r3866pua, tribro-mophosphine
PubChem CID 24614
IUPAC Name tribromophosphane
SMILES P(Br)(Br)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC169471000
SDS View Documents
Thermo Scientific Chemicals
169471000
100 g Glass bottle
Each for $116.26
 
AC169475000
SDS View Documents
Thermo Scientific Chemicals
169475000
500 g Glass bottle
Each for $293.74
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
Specifications

Chemical Identifiers

7789-60-8
270.69
IPNPIHIZVLFAFP-UHFFFAOYSA-N
24614
P(Br)(Br)Br
Br3P
MFCD00011436
phosphorus tribromide, phosphorous tribromide, tribromophosphine, phosphorus iii bromide, phosphorous bromide, pbr3, phosphorus bromide, phosphorus bromide pbr3, unii-58r3866pua, tribro-mophosphine
tribromophosphane

Specifications

175.0°C
7789-60-8
Liquid
98.5% min. (ex Br) (Argentometry)
PBr3
01,873; 02,330; 07,292
phosphorus tribromide, phosphorous tribromide, tribromophosphine, phosphorus iii bromide, phosphorous bromide, pbr3, phosphorus bromide, phosphorus bromide pbr3, unii-58r3866pua, tribro-mophosphine
IPNPIHIZVLFAFP-UHFFFAOYSA-N
tribromophosphane
24614
99%
1.697
2.8800g/mL
-40.0°C
Brown or Colorless to Yellow
100 g
Br3P
MFCD00011436
15, 7469
Solubility in water: reacts. Other solubilities: soluble in acetone,alcohol,carbon disulfide,,hydrogen sulfide,methanol,ether,chloroform and,carbon tetrachloride
P(Br)(Br)Br
270.69
270.69
Glass bottle
2.88
Phosphorus tribromide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Reacts violently with water.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 232-178-2

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only