missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phthalic acid, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 131070025

 View more versions of this product

Catalog No. AC131070025



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Phthalic acid
88-99-3
99%
MFCD00002467
15, 7483
Solubility in water: 7g/L (25°C). Other solubilities: soluble in alcohol, sparingly soluble in ether, practically insoluble in chloroform
C1=CC=C(C(=C1)C(=O)O)C(=O)O
166.132
CHEBI:29069
99%
2.5 kg
Authentic
Fine Crystalline Powder
200.0°C to 205.0°C
White
C8H6O4
09, 791
1,2-benzenedicarboxylic acid, benzene-1,2-dicarboxylic acid, o-phthalic acid, o-dicarboxybenzene, o-benzenedicarboxylic acid, pathalic acid, acide phtalique, kyselina ftalova, ortho-phthalic acid, phthalicacid
XNGIFLGASWRNHJ-UHFFFAOYSA-N
phthalic acid
1017
166.13
168g/mL
168°C
Plastic bottle

Chemical Identifiers

88-99-3
166.132
XNGIFLGASWRNHJ-UHFFFAOYSA-N
1017
phthalic acid
C8H6O4
MFCD00002467
1,2-benzenedicarboxylic acid, benzene-1,2-dicarboxylic acid, o-phthalic acid, o-dicarboxybenzene, o-benzenedicarboxylic acid, pathalic acid, acide phtalique, kyselina ftalova, ortho-phthalic acid, phthalicacid
CHEBI:29069
C1=CC=C(C(=C1)C(=O)O)C(=O)O
Safety and Handling

Safety and Handling

GHS P Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only