Learn More
Phthalic acid, 99%, Thermo Scientific Chemicals
$154.69 - $516.09
Chemical Identifiers
CAS | 88-99-3 |
---|---|
Molecular Formula | C8H6O4 |
Molecular Weight (g/mol) | 166.132 |
MDL Number | MFCD00002467 |
InChI Key | XNGIFLGASWRNHJ-UHFFFAOYSA-N |
Synonym | 1,2-benzenedicarboxylic acid, benzene-1,2-dicarboxylic acid, o-phthalic acid, o-dicarboxybenzene, o-benzenedicarboxylic acid, pathalic acid, acide phtalique, kyselina ftalova, ortho-phthalic acid, phthalicacid |
PubChem CID | 1017 |
ChEBI | CHEBI:29069 |
IUPAC Name | phthalic acid |
SMILES | C1=CC=C(C(=C1)C(=O)O)C(=O)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC131072500
|
Thermo Scientific Chemicals
131072500 |
250 g | Plastic bottle |
Each for $154.69
|
|
||||
AC131070010
|
Thermo Scientific Chemicals
131070010 |
1 kg | Plastic bottle |
Each for $227.01
|
|
||||
AC131070025
|
Thermo Scientific Chemicals
131070025 |
2.5 kg | Plastic bottle |
Each for $516.09
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
88-99-3 | |
166.132 | |
XNGIFLGASWRNHJ-UHFFFAOYSA-N | |
1017 | |
phthalic acid |
C8H6O4 | |
MFCD00002467 | |
1,2-benzenedicarboxylic acid, benzene-1,2-dicarboxylic acid, o-phthalic acid, o-dicarboxybenzene, o-benzenedicarboxylic acid, pathalic acid, acide phtalique, kyselina ftalova, ortho-phthalic acid, phthalicacid | |
CHEBI:29069 | |
C1=CC=C(C(=C1)C(=O)O)C(=O)O |
Specifications
200.0°C to 205.0°C | |
White | |
C8H6O4 | |
09, 791 | |
1,2-benzenedicarboxylic acid, benzene-1,2-dicarboxylic acid, o-phthalic acid, o-dicarboxybenzene, o-benzenedicarboxylic acid, pathalic acid, acide phtalique, kyselina ftalova, ortho-phthalic acid, phthalicacid | |
XNGIFLGASWRNHJ-UHFFFAOYSA-N | |
phthalic acid | |
1017 | |
166.13 | |
168g/mL | |
168°C | |
Plastic bottle | |
Phthalic acid |
88-99-3 | |
99% | |
MFCD00002467 | |
15, 7483 | |
Solubility in water: 7g/L (25°C). Other solubilities: soluble in alcohol, sparingly soluble in ether, practically insoluble in chloroform | |
C1=CC=C(C(=C1)C(=O)O)C(=O)O | |
166.132 | |
CHEBI:29069 | |
99% | |
250 g | |
Authentic | |
Fine Crystalline Powder |
Safety and Handling
GHS P Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
RUO – Research Use Only