Learn More
Phthalic acid, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 131072500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Phthalic acid | |
88-99-3 | |
99% | |
MFCD00002467 | |
15, 7483 | |
Solubility in water: 7g/L (25°C). Other solubilities: soluble in alcohol, sparingly soluble in ether, practically insoluble in chloroform | |
C1=CC=C(C(=C1)C(=O)O)C(=O)O | |
166.132 | |
CHEBI:29069 | |
99% | |
250 g | |
Authentic | |
Fine Crystalline Powder |
200.0°C to 205.0°C | |
White | |
C8H6O4 | |
09, 791 | |
1,2-benzenedicarboxylic acid, benzene-1,2-dicarboxylic acid, o-phthalic acid, o-dicarboxybenzene, o-benzenedicarboxylic acid, pathalic acid, acide phtalique, kyselina ftalova, ortho-phthalic acid, phthalicacid | |
XNGIFLGASWRNHJ-UHFFFAOYSA-N | |
phthalic acid | |
1017 | |
166.13 | |
168g/mL | |
168°C | |
Plastic bottle |
Chemical Identifiers
88-99-3 | |
166.132 | |
XNGIFLGASWRNHJ-UHFFFAOYSA-N | |
1017 | |
phthalic acid |
C8H6O4 | |
MFCD00002467 | |
1,2-benzenedicarboxylic acid, benzene-1,2-dicarboxylic acid, o-phthalic acid, o-dicarboxybenzene, o-benzenedicarboxylic acid, pathalic acid, acide phtalique, kyselina ftalova, ortho-phthalic acid, phthalicacid | |
CHEBI:29069 | |
C1=CC=C(C(=C1)C(=O)O)C(=O)O |
Safety and Handling
GHS P Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
RUO – Research Use Only