missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phthalimidoacetaldehyde diethyl acetal, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1849106

 View more versions of this product

Catalog No. AAA1849106


Explore available promotions

Description

Description

Phthalimidoacetaldehyde diethyl acetal is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phthalimidoacetaldehyde diethyl acetal is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Specifications

Phthalimidoacetaldehyde diethyl acetal
71°C to 74°C
MFCD00005901
215793
Insoluble in water.
CCOC(CN1C(=O)C2=CC=CC=C2C1=O)OCC
263.29
263.3
78902-09-7
C14H17NO4
5 g
phthalimidoacetaldehyde diethyl acetal, 2-2,2-diethoxyethyl isoindoline-1,3-dione, n-2,2-diethoxyethyl phthalimide, 2-phthalimidoacetaldehyde diethyl acetal, 2-phthalimido acetaldehyde diethylacetal, 2-2,2-diethoxyethyl isoindole-1,3-dione, 1h-isoindole-1,3 2h-dione, 2-2,2-diethoxyethyl, 2-2,2-diethoxyethyl-1h-isoindole-1,3 2h-dione, 2-2,2-diethoxyethyl benzo c azoline-1,3-dione, acmc-209pfw
GEFXJJJQUSEHLV-UHFFFAOYSA-N
2-(2,2-diethoxyethyl)isoindole-1,3-dione
315286
99%

Chemical Identifiers

78902-09-7
263.29
GEFXJJJQUSEHLV-UHFFFAOYSA-N
315286
CCOC(CN1C(=O)C2=CC=CC=C2C1=O)OCC
C14H17NO4
MFCD00005901
phthalimidoacetaldehyde diethyl acetal, 2-2,2-diethoxyethyl isoindoline-1,3-dione, n-2,2-diethoxyethyl phthalimide, 2-phthalimidoacetaldehyde diethyl acetal, 2-phthalimido acetaldehyde diethylacetal, 2-2,2-diethoxyethyl isoindole-1,3-dione, 1h-isoindole-1,3 2h-dione, 2-2,2-diethoxyethyl, 2-2,2-diethoxyethyl-1h-isoindole-1,3 2h-dione, 2-2,2-diethoxyethyl benzo c azoline-1,3-dione, acmc-209pfw
2-(2,2-diethoxyethyl)isoindole-1,3-dione
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only