missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phthalimidoacetaldehyde diethyl acetal, 99%, Thermo Scientific Chemicals

$71.58 - $216.90

Chemical Identifiers

CAS 78902-09-7
Molecular Formula C14H17NO4
Molecular Weight (g/mol) 263.29
MDL Number MFCD00005901
InChI Key GEFXJJJQUSEHLV-UHFFFAOYSA-N
Synonym phthalimidoacetaldehyde diethyl acetal, 2-2,2-diethoxyethyl isoindoline-1,3-dione, n-2,2-diethoxyethyl phthalimide, 2-phthalimidoacetaldehyde diethyl acetal, 2-phthalimido acetaldehyde diethylacetal, 2-2,2-diethoxyethyl isoindole-1,3-dione, 1h-isoindole-1,3 2h-dione, 2-2,2-diethoxyethyl, 2-2,2-diethoxyethyl-1h-isoindole-1,3 2h-dione, 2-2,2-diethoxyethyl benzo c azoline-1,3-dione, acmc-209pfw
PubChem CID 315286
IUPAC Name 2-(2,2-diethoxyethyl)isoindole-1,3-dione
SMILES CCOC(CN1C(=O)C2=CC=CC=C2C1=O)OCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1849106
View Documents
Thermo Scientific Chemicals
A1849106
5 g
Each for $71.58
 
AAA1849114
View Documents
Thermo Scientific Chemicals
A1849114
25 g
Each for $216.90
 
Description

Description

Phthalimidoacetaldehyde diethyl acetal is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phthalimidoacetaldehyde diethyl acetal is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

78902-09-7
263.29
GEFXJJJQUSEHLV-UHFFFAOYSA-N
315286
CCOC(CN1C(=O)C2=CC=CC=C2C1=O)OCC
C14H17NO4
MFCD00005901
phthalimidoacetaldehyde diethyl acetal, 2-2,2-diethoxyethyl isoindoline-1,3-dione, n-2,2-diethoxyethyl phthalimide, 2-phthalimidoacetaldehyde diethyl acetal, 2-phthalimido acetaldehyde diethylacetal, 2-2,2-diethoxyethyl isoindole-1,3-dione, 1h-isoindole-1,3 2h-dione, 2-2,2-diethoxyethyl, 2-2,2-diethoxyethyl-1h-isoindole-1,3 2h-dione, 2-2,2-diethoxyethyl benzo c azoline-1,3-dione, acmc-209pfw
2-(2,2-diethoxyethyl)isoindole-1,3-dione

Specifications

78902-09-7
C14H17NO4
5 g
phthalimidoacetaldehyde diethyl acetal, 2-2,2-diethoxyethyl isoindoline-1,3-dione, n-2,2-diethoxyethyl phthalimide, 2-phthalimidoacetaldehyde diethyl acetal, 2-phthalimido acetaldehyde diethylacetal, 2-2,2-diethoxyethyl isoindole-1,3-dione, 1h-isoindole-1,3 2h-dione, 2-2,2-diethoxyethyl, 2-2,2-diethoxyethyl-1h-isoindole-1,3 2h-dione, 2-2,2-diethoxyethyl benzo c azoline-1,3-dione, acmc-209pfw
GEFXJJJQUSEHLV-UHFFFAOYSA-N
2-(2,2-diethoxyethyl)isoindole-1,3-dione
315286
99%
71°C to 74°C
MFCD00005901
215793
Insoluble in water.
CCOC(CN1C(=O)C2=CC=CC=C2C1=O)OCC
263.29
263.3
Phthalimidoacetaldehyde diethyl acetal
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only