missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Phthalimidoacetaldehyde diethyl acetal, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1849114
Description
Phthalimidoacetaldehyde diethyl acetal is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPhthalimidoacetaldehyde diethyl acetal is used as a pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
Phthalimidoacetaldehyde diethyl acetal | |
71°C to 74°C | |
MFCD00005901 | |
215793 | |
Insoluble in water. | |
CCOC(CN1C(=O)C2=CC=CC=C2C1=O)OCC | |
263.29 | |
263.3 |
78902-09-7 | |
C14H17NO4 | |
25 g | |
phthalimidoacetaldehyde diethyl acetal, 2-2,2-diethoxyethyl isoindoline-1,3-dione, n-2,2-diethoxyethyl phthalimide, 2-phthalimidoacetaldehyde diethyl acetal, 2-phthalimido acetaldehyde diethylacetal, 2-2,2-diethoxyethyl isoindole-1,3-dione, 1h-isoindole-1,3 2h-dione, 2-2,2-diethoxyethyl, 2-2,2-diethoxyethyl-1h-isoindole-1,3 2h-dione, 2-2,2-diethoxyethyl benzo c azoline-1,3-dione, acmc-209pfw | |
GEFXJJJQUSEHLV-UHFFFAOYSA-N | |
2-(2,2-diethoxyethyl)isoindole-1,3-dione | |
315286 | |
99% |
Chemical Identifiers
78902-09-7 | |
263.29 | |
GEFXJJJQUSEHLV-UHFFFAOYSA-N | |
315286 | |
CCOC(CN1C(=O)C2=CC=CC=C2C1=O)OCC |
C14H17NO4 | |
MFCD00005901 | |
phthalimidoacetaldehyde diethyl acetal, 2-2,2-diethoxyethyl isoindoline-1,3-dione, n-2,2-diethoxyethyl phthalimide, 2-phthalimidoacetaldehyde diethyl acetal, 2-phthalimido acetaldehyde diethylacetal, 2-2,2-diethoxyethyl isoindole-1,3-dione, 1h-isoindole-1,3 2h-dione, 2-2,2-diethoxyethyl, 2-2,2-diethoxyethyl-1h-isoindole-1,3 2h-dione, 2-2,2-diethoxyethyl benzo c azoline-1,3-dione, acmc-209pfw | |
2-(2,2-diethoxyethyl)isoindole-1,3-dione |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only