missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phytosphingosine 98.0+%, TCI America™

$159.17 - $477.51

Chemical Identifiers

CAS 554-62-1
Molecular Formula C18H39NO3
Molecular Weight (g/mol) 317.514
MDL Number MFCD00079232
InChI Key AERBNCYCJBRYDG-KSZLIROESA-N
Synonym (2S,3S,4R)-2-Amino-1,3,4-octadecanetriol
PubChem CID 122121
ChEBI CHEBI:46961
IUPAC Name (2S,3S,4R)-2-aminooctadecane-1,3,4-triol
SMILES CCCCCCCCCCCCCCC(C(C(CO)N)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
P17651G
View Documents
TCI America
P17651G
1 g N/A
 
P17655G
View Documents
TCI America
P17655G
5 g N/A
 
Description

Description

Specifications

Chemical Identifiers

554-62-1
317.514
AERBNCYCJBRYDG-KSZLIROESA-N
122121
(2S,3S,4R)-2-aminooctadecane-1,3,4-triol
C18H39NO3
MFCD00079232
(2S,3S,4R)-2-Amino-1,3,4-octadecanetriol
CHEBI:46961
CCCCCCCCCCCCCCC(C(C(CO)N)O)O

Specifications

554-62-1
C18H39NO3
1 g
AERBNCYCJBRYDG-KSZLIROESA-N
(2S,3S,4R)-2-aminooctadecane-1,3,4-triol
122121
317.51
Crystalline Powder
102°C
MFCD00079232
(2S,3S,4R)-2-Amino-1,3,4-octadecanetriol
CCCCCCCCCCCCCCC(C(C(CO)N)O)O
317.514
CHEBI:46961
≥98.0% (T)
Phytosphingosine
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'