missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phytosphingosine 98.0+%, TCI America™

Supplier:  TCI America P17651G

 View more versions of this product

Catalog No. P17651G


Explore available promotions

Description

Description

Specifications

Specifications

Phytosphingosine
102°C
MFCD00079232
(2S,3S,4R)-2-Amino-1,3,4-octadecanetriol
CCCCCCCCCCCCCCC(C(C(CO)N)O)O
317.514
CHEBI:46961
≥98.0% (T)
554-62-1
C18H39NO3
1 g
AERBNCYCJBRYDG-KSZLIROESA-N
(2S,3S,4R)-2-aminooctadecane-1,3,4-triol
122121
317.51
Crystalline Powder

Chemical Identifiers

554-62-1
317.514
AERBNCYCJBRYDG-KSZLIROESA-N
122121
(2S,3S,4R)-2-aminooctadecane-1,3,4-triol
C18H39NO3
MFCD00079232
(2S,3S,4R)-2-Amino-1,3,4-octadecanetriol
CHEBI:46961
CCCCCCCCCCCCCCC(C(C(CO)N)O)O
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions