Learn More
Thermo Scientific™ Acetonitrile (ACN), LC-MS Grade
An LC/MS-grade preparation with high purity and low UV-absorptivity that makes it suitable for HPLC and mass spectrometry applications.
$258.39 - $258.39
Specifications
Grade | LC-MS |
---|---|
Item Name | Acetonitrile (ACN) |
Content And Storage | Store between 2 to 25°C in a dry, cool, well-ventilated area, away from incompatible materials. |
Form | Liquid |
Reagent Type | Solvent (Pure) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
PI51101
|
Thermo Scientific™
T51101K2 |
1 L |
Each for $258.39
|
|
||||||
Description
Thermo Scientific™ Acetonitrile (ACN) is an LC/MS-grade preparation with high purity and low UV-absorptivity that makes it suitable for HPLC and mass spectrometry applications.
Thermo Scientific LC/MS Grade Acetonitrile is specially purified by a proprietary method and tested to ensure lot-to-lot consistency with a low UV absorbance to provide you with the most sensitive detection across all wavelengths. Acetonitrile is 0.2-micron filtered, packaged in solvent-rinsed amber glass bottles and sealed under a nitrogen atmosphere with PTFE-lined fluorocarbon caps for ultimate protection.
Highlights:
- 37 quality tests ensure low, stable baselines and lot-to-lot consistency
- Low UV absorbance yields low baselines and high detection sensitivity
- Low impurities protects columns and simplifies analysis by eliminating extraneous peaks
- Specially purified and tested to the highest specifications to ensure data integrity, maximize assay sensitivity, and to prolong equipment life
- Store between 2°C to 25°C in a dry, cool, well-ventilated area, away from incompatible materials
Product Details:
- Alternative names: Methyl cyanide, cyanomethane, ethanenitrile
- Molecular formula: CH3CN
- Molecular weight: 41.05
- Density: 0.780 g/mL
Specifications
LC-MS | |
Store between 2 to 25°C in a dry, cool, well-ventilated area, away from incompatible materials. | |
Solvent (Pure) | |
HPLC, Mass Spectrometry |
Acetonitrile (ACN) | |
Liquid | |
Pierce™ | |
75-05-8 |
For Research Use Only. Not for use in diagnostic procedures.