missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Acetonitrile (ACN), LC-MS Grade

An LC/MS-grade preparation with high purity and low UV-absorptivity that makes it suitable for HPLC and mass spectrometry applications.

Supplier:  Thermo Scientific™ T51101K2

 View more versions of this product

Catalog No. PI51101

Explore available promotions

Description

Description

Thermo Scientific™ Acetonitrile (ACN) is an LC/MS-grade preparation with high purity and low UV-absorptivity that makes it suitable for HPLC and mass spectrometry applications.

Thermo Scientific LC/MS Grade Acetonitrile is specially purified by a proprietary method and tested to ensure lot-to-lot consistency with a low UV absorbance to provide you with the most sensitive detection across all wavelengths. Acetonitrile is 0.2-micron filtered, packaged in solvent-rinsed amber glass bottles and sealed under a nitrogen atmosphere with PTFE-lined fluorocarbon caps for ultimate protection.

Highlights:

  • 37 quality tests ensure low, stable baselines and lot-to-lot consistency
  • Low UV absorbance yields low baselines and high detection sensitivity
  • Low impurities protects columns and simplifies analysis by eliminating extraneous peaks
  • Specially purified and tested to the highest specifications to ensure data integrity, maximize assay sensitivity, and to prolong equipment life
  • Store between 2°C to 25°C in a dry, cool, well-ventilated area, away from incompatible materials

Product Details:

  • Alternative names: Methyl cyanide, cyanomethane, ethanenitrile
  • Molecular formula: CH3CN
  • Molecular weight: 41.05
  • Density: 0.780 g/mL
Specifications

Specifications

LC-MS
1 L
Liquid
Pierce™
75-05-8
Acetonitrile (ACN)
Store between 2 to 25°C in a dry, cool, well-ventilated area, away from incompatible materials.
Solvent (Pure)
HPLC, Mass Spectrometry
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.