missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ ECL Western Blotting Substrate

Value-priced, entry-level peroxidase substrate for enhanced chemiluminescence (ECL) that directly replaces costlier products without the need to re-optimize conditions.

$166.21 - $1688.29

Specifications

Product Type Substrate
Detection Method Chemiluminescence
Form Liquid
Product Line Pierce
Substrate Horseradish Peroxidase
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PI32109
View Documents
Thermo Scientific™
LSG32109
50 mL
Each for $166.21
Only null left
 
FER32209X4
View Documents
Thermo Scientific™
32209X4
4 x 250 mL
Each for $1,278.46
Only null left
 
PI32209
View Documents
Thermo Scientific™
32209
250 mL
Each for $416.86
Only null left
 
FER32106X4
View Documents
Thermo Scientific™
32106X4
4 x 500 mL
Each for $1,688.29
Only null left
 
PI32106
View Documents
Thermo Scientific™
32106
500 mL
Each for $639.72
Only null left
 
Description

Description

Thermo Scientific™ Pierce ECL Western Blotting Substrate is a value-priced, entry-level peroxidase substrate for enhanced chemiluminescence (ECL) that directly replaces costlier products without the need to re-optimize conditions.

Pierce ECL Western Blotting Substrate provides reliability and performance equivalent to other standard enhanced chemilumescent (ECL) substrates for detection of horseradish peroxidase (HRP) enzyme activity, yet costs much less. Because the luminol and peroxide reagent-formulations are identical to other commercially available substrate products, one can switch to Pierce ECL without needing to optimize probing conditions or incubation protocols. Simply switch to Pierce ECL and save a bundle.

Highlights:

  • ECL substrate – an enhanced chemiluminescent substrate for detection of horseradish peroxidase (HRP) activity from antibodies and other Western blot probes

  • About half the cost of other ECL Substrates – low overhead and a commitment to customer value enables Thermo Fisher Scientific to make this product available to you at a significantly lower price than other companies (these claims are based on the 2010 U.S. list prices)

  • No optimization required – switching to Pierce ECL substrate from other entry-level ECL substrates does not require optimization or protocol changes

  • A name you can can rely on – count on the strong quality assurance, technical support and reputation behind Thermo Scientific Pierce Protein Research Products

Includes:

Detection Reagent 1 and 2

Specifications

Specifications

Substrate
Liquid
Horseradish Peroxidase
Upon receipt store at 2-8°C. Product is shipped at ambient temperature.
Chemiluminescence
Pierce
1 to 2 hr.
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

For Research Use Only. Not for use in diagnostic procedures.