Learn More
Thermo Scientific™ Pierce™ Prestained Protein MW Marker
Thermo Scientific Prestained Protein Molecular Weight Marker is a mixture of six blue-stained natural proteins (20 to 120 kDa) for use as size standards in protein electrophoresis (SDS-PAGE).
$148.39 - $148.39
Specifications
Content And Storage | Storage Buffer: 62.5 mM Tris · H3PO4 (pH 7.5 at 25°C), 1 mM EDTA, 2% (w/v) SDS, 1 mM DTT, 1.5 mM NaN3 and 33% (v/v) glycerol. Upon receipt store at -20°C. Product is shipped with an ice pack. |
---|---|
Detection Method | Colorimetric |
Number of Markers | 6 |
Product Type | Protein Molecular Weight (MW) Marker |
Ready to Load | Yes |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
PI26612
|
Thermo Scientific™
26612 |
500 μL |
Each for $148.39
|
|
|||||
Description
Thermo Scientific™ Prestained Protein Molecular Weight Marker is a mixture of six blue-stained natural proteins (20 to 120kDa) for use as size standards in protein electrophoresis (SDS-PAGE).
Thermo Scientific Prestained Protein Molecular Weight Marker is a mixture of six blue-stained natural proteins (20 to 120 kDa) for use as size standards in protein electrophoresis (SDS-PAGE). The protein standards are supplied in a ready-to-use format for direct loading onto gels; no need to heat, reduce, or add sample buffer prior to use.
Component proteins
- Beta-galactosidase (120 kDa)
- Bovine serum albumin (85 kDa)
- Ovalbumin (50 kDa)
- Cardonic anhydrase (35 kDa)
- Beta-lactoglobulin (25 kDa)
- Lysozyme (20 kDa)
Applications
- Monitoring protein migration during SDS-polyacrylamide gel electrophoresis.
- Monitoring protein transfer onto membranes after western blotting
- Sizing of proteins on SDS-polyacrylamide gels and western blots
Specifications
Storage Buffer: 62.5 mM Tris · H3PO4 (pH 7.5 at 25°C), 1 mM EDTA, 2% (w/v) SDS, 1 mM DTT, 1.5 mM NaN3 and 33% (v/v) glycerol. Upon receipt store at -20°C. Product is shipped with an ice pack. |
|
6 | |
Yes | |
1 color: Blue | |
Pierce™ |
Colorimetric | |
Protein Molecular Weight (MW) Marker | |
20 to 120 kDa | |
120, 85, 50, 35, 25, 20 kDa |
For Research Use Only. Not for use in diagnostic procedures.