missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Pierce™ Silver Stain Kit

Stain proteins in 1D and 2D gels with this rapid, ultra-sensitive and versatile silver stain system that yields consistent and reliable results.

Supplier:  Thermo Scientific™ 24612

 View more versions of this product

Catalog No. PI24612


Explore available promotions

Description

Description

Thermo Scientific™ Pierce Silver Stain Kit is a rapid, ultra-sensitive and versatile silver stain system for protein detection in polyacrylamide gels, yielding consistent and reliable results with many 1D and 2D gel types for both first-time and experienced users.

The Pierce Silver Stain Kit is a rapid, ultra-sensitive, and versatile silver stain system for protein detection in polyacrylamide gels, yielding consistent and reliable results. It is a metallic silver (Ag) protein stain that yields a remarkably clear and uniform gel background. In standard mini gels, proteins are detectable at greater than 0.25 ng per band or spot. The protocol has been optimized for flexibility by allowing short or overnight gel fixation and staining steps without affecting staining performance (sensitivity or clarity). A short (one-minute) sensitization step, performed after gel fixation, yields results that are free of the characteristically dark or blotchy backgrounds often seen with homemade or other commercially available silver stains. This feature is beneficial when performing densitometric analysis of silver stained gels.

Features:
Sensitivity—detect proteins at 0.25 ng
Protocol—gel staining can be performed within 5 min to 20 hrs, depending on desired sensitivity
Gel-compatibility—staining performance is very good with a variety of commercial precast mini-gels and SDS-PAGE buffer-systems
Workflow-compatible—mild chemical formulation ensures compatibility with mass spectrometry, sequencing, and other downstream procedures that depend on destaining and protein recovery

Silver staining methods generally use either glutaraldehyde or formaldehyde, which cause some covalent crosslinking of protein to each other and the gel matrix. To the extent that this crosslinking occurs, extraction or elution of protein from the gel will be inhibited. Pierce Silver Stain uses formaldehyde in the stain and developer working solutions, but the concentration is low enough that irreversible protein crosslinking is minimized.

Specifications

Specifications

Sufficient For: 20 mini gels (SDS-PAGE)
• Silver Stain Sensitizer, 2 mL
• Silver Stain, 500 mL
• Silver Stain Enhancer, 25 mL
• Silver Stain Developer, 500 mL

Store at room temperature.
Colorimetric
Stain Kit
1 L
In-Gel Detection
Silver
Pierce™
Protein
Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.