missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Pinacol, 99%, Thermo Scientific Chemicals

Pinacol, 99%, C6H14O2, CAS Number-76-09-5, pinacone, pinacol, unii-527qe7i5co, tetramethylethylene glycol, 2,3-dimethyl-2,3-butanediol, 2,3-dimethyl-2,3-dihydroxybutane, 1,1,2,2-tetramethylethylene glycol, 2,3-butanediol, 2,3-dimethyl, 2,3-dihydroxy-2,3-dimethylbutane, 2,3-dimethyl-butane-2,3-diol

Supplier:  Thermo Scientific Chemicals 418001000

 View more versions of this product

Catalog No. AC418001000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Pinacol
76-09-5
White
77°C
98.5% min. (GC)
C6H14O2
MFCD00004462
01,487
pinacol, 2,3-dimethyl-2,3-butanediol, pinacone, 2,3-butanediol, 2,3-dimethyl, tetramethylethylene glycol, 1,1,2,2-tetramethylethylene glycol, unii-527qe7i5co, 2,3-dimethyl-butane-2,3-diol, 2,3-dihydroxy-2,3-dimethylbutane, 2,3-dimethyl-2,3-dihydroxybutane
IVDFJHOHABJVEH-UHFFFAOYSA-N
2,3-dimethylbutane-2,3-diol
6425
99%
99%
40.0°C to 43.0°C
174.0°C
Authentic
Glass bottle
(CH3)2COHCOH(CH3)2
100 g
15,755
Solubility in water: insoluble
CC(C)(C(C)(C)O)O
118.18
118.18
Crystalline Low Melting Solid

Chemical Identifiers

76-09-5
118.18
IVDFJHOHABJVEH-UHFFFAOYSA-N
6425
CC(C)(C(C)(C)O)O
C6H14O2
MFCD00004462
pinacol, 2,3-dimethyl-2,3-butanediol, pinacone, 2,3-butanediol, 2,3-dimethyl, tetramethylethylene glycol, 1,1,2,2-tetramethylethylene glycol, unii-527qe7i5co, 2,3-dimethyl-butane-2,3-diol, 2,3-dihydroxy-2,3-dimethylbutane, 2,3-dimethyl-2,3-dihydroxybutane
2,3-dimethylbutane-2,3-diol
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 200-933-5

RTECSNumber : EK1720000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only