Learn More
Pinacol, 99%, Thermo Scientific Chemicals
Pinacol, 99%, C6H14O2, CAS Number-76-09-5, pinacone, pinacol, unii-527qe7i5co, tetramethylethylene glycol, 2,3-dimethyl-2,3-butanediol, 2,3-dimethyl-2,3-dihydroxybutane, 1,1,2,2-tetramethylethylene glycol, 2,3-butanediol, 2,3-dimethyl, 2,3-dihydroxy-2,3-dimethylbutane, 2,3-dimethyl-butane-2,3-diol
$212.37 - $1744.21
Chemical Identifiers
CAS | 76-09-5 |
---|---|
Molecular Formula | C6H14O2 |
Molecular Weight (g/mol) | 118.18 |
MDL Number | MFCD00004462 |
InChI Key | IVDFJHOHABJVEH-UHFFFAOYSA-N |
Synonym | pinacol, 2,3-dimethyl-2,3-butanediol, pinacone, 2,3-butanediol, 2,3-dimethyl, tetramethylethylene glycol, 1,1,2,2-tetramethylethylene glycol, unii-527qe7i5co, 2,3-dimethyl-butane-2,3-diol, 2,3-dihydroxy-2,3-dimethylbutane, 2,3-dimethyl-2,3-dihydroxybutane |
PubChem CID | 6425 |
IUPAC Name | 2,3-dimethylbutane-2,3-diol |
SMILES | CC(C)(C(C)(C)O)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC418001000
|
Thermo Scientific Chemicals
418001000 |
100 g | Glass bottle |
Each for $212.37
|
|
||||
AC418002500
|
Thermo Scientific Chemicals
418002500 |
250 g | Glass bottle |
Each for $477.27
|
|
||||
AC418000010
|
Thermo Scientific Chemicals
418000010 |
1 kg | Glass bottle |
Each for $1,744.21
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
76-09-5 | |
118.18 | |
IVDFJHOHABJVEH-UHFFFAOYSA-N | |
6425 | |
CC(C)(C(C)(C)O)O |
C6H14O2 | |
MFCD00004462 | |
pinacol, 2,3-dimethyl-2,3-butanediol, pinacone, 2,3-butanediol, 2,3-dimethyl, tetramethylethylene glycol, 1,1,2,2-tetramethylethylene glycol, unii-527qe7i5co, 2,3-dimethyl-butane-2,3-diol, 2,3-dihydroxy-2,3-dimethylbutane, 2,3-dimethyl-2,3-dihydroxybutane | |
2,3-dimethylbutane-2,3-diol |
Specifications
76-09-5 | |
White | |
77°C | |
98.5% min. (GC) | |
C6H14O2 | |
MFCD00004462 | |
01,487 | |
pinacol, 2,3-dimethyl-2,3-butanediol, pinacone, 2,3-butanediol, 2,3-dimethyl, tetramethylethylene glycol, 1,1,2,2-tetramethylethylene glycol, unii-527qe7i5co, 2,3-dimethyl-butane-2,3-diol, 2,3-dihydroxy-2,3-dimethylbutane, 2,3-dimethyl-2,3-dihydroxybutane | |
IVDFJHOHABJVEH-UHFFFAOYSA-N | |
2,3-dimethylbutane-2,3-diol | |
6425 | |
99% | |
Pinacol, 99% |
40.0°C to 43.0°C | |
174.0°C | |
Authentic | |
Glass bottle | |
(CH3)2COHCOH(CH3)2 | |
100 g | |
15,755 | |
Solubility in water: insoluble | |
CC(C)(C(C)(C)O)O | |
118.18 | |
118.18 | |
Crystalline Low Melting Solid |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 200-933-5
RTECSNumber : EK1720000
TSCA : TSCA
RUO – Research Use Only