OHAUS™ Pioneer PX Precision Balances
OHAUS Corporation™ Pioneer PX Precision Balance offers high accuracy and repeatability for applications in laboratory, industrial and education settings. Features a second line display for additional information and connectivity for easy communication
$1754.00 - $3307.93
Specifications
Certifications/Compliance | Electromagnetic Compatibility: IEC/EN 61326-1 Class B, Basic Environments; FCC Part 15 Class A; Canada ICES-003 Class A, CSA |
---|---|
Connection Type | RS232, USB |
Display Type | 2 Line Backlit LCD |
Leveling | Integrated level bubble |
Linearity | ±2 Divisions |
Includes: AC Power Adapter
Catalog Number | Mfr. No. | Capacity | Readability | Type | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity | Readability | Type | Price | Quantity | |||
01922178
|
OHAUS™
30429848 |
160 g | 0.001 g | External Calibration |
Each for $1,957.07
|
|
|||
01922180
|
OHAUS™
30429849 |
320 g | 0.001 g | External Calibration |
Each for $2,280.20
|
|
|||
01922179
|
OHAUS™
OHS30429839 |
160 g | 0.001 g | Internal Calibration |
Each for $2,437.63
|
|
|||
01922181
|
OHAUS™
OHS30429840 |
320 g | 0.001 g | Internal Calibration |
Each for $2,744.02
|
|
|||
01922190
|
OHAUS™
30430058 |
1600 g | 0.01 g | External Calibration |
Each for $1,888.98
|
|
|||
01922192
|
OHAUS™
30429850 |
2200 g | 0.01 g | External Calibration |
Each for $2,064.07
|
|
|||
01922194
|
OHAUS™
30429851 |
3200 g | 0.01 g | External Calibration |
Each for $2,497.13
|
|
|||
01922184
|
OHAUS™
30429852 |
4200 g | 0.01 g | External Calibration |
Each for $2,820.80
|
|
|||
01922186
|
OHAUS™
30430059 |
2200 g | 0.1 g | External Calibration |
Each for $1,754.00
|
|
|||
01922188
|
OHAUS™
30429853 |
4200 g | 0.1 g | External Calibration |
Each for $2,025.43
|
|
|||
01922189
|
OHAUS™
30429844 |
4200 g | 0.1 g | Internal Calibration |
Each for $2,428.50
|
|
|||
01922191
|
OHAUS™
30430055 |
1600 g | 0.01 g | Internal Calibration |
Each for $2,309.82
|
|
|||
01922193
|
OHAUS™
30429841 |
2200 g | 0.01 g | Internal Calibration |
Each for $2,496.91
|
|
|||
01922195
|
OHAUS™
30429842 |
3200 g | 0.01 g | Internal Calibration |
Each for $2,948.91
|
|
|||
01922185
|
OHAUS™
30429843 |
4200 g | 0.01 g | Internal Calibration |
Each for $3,307.93
|
|
|||
01922187
|
OHAUS™
30430056 |
2200 g | 0.1 g | Internal Calibration |
Each for $2,145.50
|
|
|||
Description
- 2 Line Display
- Durable Construction
- Communication Ports (RS-232, USB)
Compliance
Electromagnetic Compatibility: IEC/EN 61326-1 Class B, Basic Environments; FCC Part 15 Class A; Canada ICES-003 Class A, CSA
Warranty and Services
2 Year
Specifications
Electromagnetic Compatibility: IEC/EN 61326-1 Class B, Basic Environments; FCC Part 15 Class A; Canada ICES-003 Class A, CSA | |
2 Line Backlit LCD | |
±2 Divisions | |
Round | |
10°C to 30°C | |
Basic Weighing, Parts Counting, Percent Weighing, Animal Weighing, Density Determination | |
209 mm | |
50/60 Hz 12/18 VA |
RS232, USB | |
Integrated level bubble | |
Cast Aluminum Base, Plastic Top Housing | |
±1 Division | |
2 Years | |
321 mm | |
Power Input: 100â240V ∼ 200mA 50â60Hz 12â18VA; Power Output: 12 VDC 0.5A |