missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Pipemidic acid, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals J6643722
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Pipemidic Acid | |
Powder | |
251°C to 255°C | |
MFCD00057291 | |
Soluble in 1N sodium hydroxide or in DMSO. Insoluble in water | |
CCN1C=C(C(O)=O)C(=O)C2=CN=C(N=C12)N1CCNCC1 | |
303.32 | |
CHEBI:75250 | |
98% | |
>110°C (230°F) | |
717°C |
Pipemidic acid | |
51940-44-4 | |
C14H17N5O3 | |
Plastic Bottle | |
JOHZPMXAZQZXHR-UHFFFAOYSA-N | |
8-ethyl-5-oxo-2-(piperazin-1-yl)-5H,8H-pyrido[2,3-d]pyrimidine-6-carboxylic acid | |
4831 | |
303.32 | |
100 g | |
Light sensitive |
Chemical Identifiers
51940-44-4 | |
303.32 | |
JOHZPMXAZQZXHR-UHFFFAOYSA-N | |
CHEBI:75250 | |
CCN1C=C(C(O)=O)C(=O)C2=CN=C(N=C12)N1CCNCC1 |
C14H17N5O3 | |
MFCD00057291 | |
4831 | |
8-ethyl-5-oxo-2-(piperazin-1-yl)-5H,8H-pyrido[2,3-d]pyrimidine-6-carboxylic acid |
Safety and Handling
P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c
H317-H334
EINECSNumber : 257-530-2
RTECSNumber : UV1153800
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only