Learn More
Pipemidic acid, 98%, Thermo Scientific Chemicals
$141.88 - $904.96
Chemical Identifiers
CAS | 51940-44-4 |
---|---|
Molecular Formula | C14H17N5O3 |
Molecular Weight (g/mol) | 303.32 |
MDL Number | MFCD00057291 |
InChI Key | JOHZPMXAZQZXHR-UHFFFAOYSA-N |
PubChem CID | 4831 |
ChEBI | CHEBI:75250 |
IUPAC Name | 8-ethyl-5-oxo-2-(piperazin-1-yl)-5H,8H-pyrido[2,3-d]pyrimidine-6-carboxylic acid |
SMILES | CCN1C=C(C(O)=O)C(=O)C2=CN=C(N=C12)N1CCNCC1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ6643709
|
Thermo Scientific Chemicals
J6643709 |
10 g |
Each for $141.88
|
|
|||||
AAJ6643722
|
Thermo Scientific Chemicals
J6643722 |
100 g |
Each for $904.96
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
51940-44-4 | |
303.32 | |
JOHZPMXAZQZXHR-UHFFFAOYSA-N | |
CHEBI:75250 | |
CCN1C=C(C(O)=O)C(=O)C2=CN=C(N=C12)N1CCNCC1 |
C14H17N5O3 | |
MFCD00057291 | |
4831 | |
8-ethyl-5-oxo-2-(piperazin-1-yl)-5H,8H-pyrido[2,3-d]pyrimidine-6-carboxylic acid |
Specifications
Pipemidic Acid | |
51940-44-4 | |
C14H17N5O3 | |
14,7460 | |
JOHZPMXAZQZXHR-UHFFFAOYSA-N | |
8-ethyl-5-oxo-2-(piperazin-1-yl)-5H,8H-pyrido[2,3-d]pyrimidine-6-carboxylic acid | |
4831 | |
303.32 | |
White | |
>110°C (230°F) | |
717°C |
Powder | |
251°C to 255°C | |
MFCD00057291 | |
Soluble in 1N sodium hydroxide or in DMSO. Insoluble in water | |
CCN1C=C(C(O)=O)C(=O)C2=CN=C(N=C12)N1CCNCC1 | |
303.32 | |
CHEBI:75250 | |
98% | |
10 g | |
Light sensitive | |
Pipemidic acid |
Safety and Handling
GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c
H317-H334
EINECSNumber : 257-530-2
RTECSNumber : UV1153800
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only