missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Poly(dimethylsiloxane), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 178445000

 View more versions of this product

Catalog No. AC178445000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Poly(dimethylsiloxane)
9016-00-6
{Si(CH3)2O-}x
96%5g/mL
Simethicone, Dimethicone, PDMS
SEUDSDUUJXTXSV-UHFFFAOYSA-N
dimethylsilanone
Authentic
Glass bottle
100mPa·s (25°C)
Oil, Free From Suspended Matter
(C2H6OSi)n
MFCD00084411
10, 8374
500 g
C[Si](C)(-*)O-*
74.15
Passes Test
1.4030 to 1.4050 (20°C, 589nm)

Chemical Identifiers

9016-00-6
74.15
SEUDSDUUJXTXSV-UHFFFAOYSA-N
dimethylsilanone
(C2H6OSi)n
MFCD00084411
Simethicone, Dimethicone, PDMS
C[Si](C)(-*)O-*
Videos
Safety and Handling

Safety and Handling

RTECSNumber : TQ2690000

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only