missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Polyethylene Glycol 1000, NF, Spectrum™ Chemical
PO118, 25322-68-3, H(OCH2CH2)nOH
Supplier: Spectrum Chemical Mfg Cor PO11845KGBL
Description
Spectrum™ Chemical Polyethylene Glycol 1000, NF acts as a lubricant, coating the surfaces in aqueous and non-aqueous environments. All Spectrum Chemical NF products, including all NF polyethylene glycol products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
4.5 to 7.5 | |
1 | |
MFCD01779601 | |
LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
ethane-1,2-diol | |
NF | |
Poly Drum |
25322-68-3 | |
(C2H4O)n | |
45 kg | |
[H]OCCO | |
62.07 | |
0.001 | |
16.0-19.0 cSt |
Chemical Identifiers
25322-68-3 | |
62.07 | |
LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
[H]OCCO |
(C2H4O)n | |
MFCD01779601 | |
ethane-1,2-diol |