missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Polyethylene Glycol 1000, NF, Spectrum™ Chemical
PO118, 25322-68-3, H(OCH2CH2)nOH
$748.26 - $5,434.79
Chemical Identifiers
CAS | 25322-68-3 |
---|---|
Molecular Formula | (C2H4O)n |
Molecular Weight (g/mol) | 62.07 |
MDL Number | MFCD01779601 |
InChI Key | LYCAIKOWRPUZTN-UHFFFAOYSA-N |
IUPAC Name | ethane-1,2-diol |
SMILES | [H]OCCO |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18605299
|
Spectrum Chemical Mfg Cor
PO11825KG |
2.5 kg | Poly Bottle |
N/A
|
|
||||
18605298
|
Spectrum Chemical Mfg Cor
PO118500GMCS6 |
6 x 500 g | Amber Glass Bottle |
N/A
|
|
||||
18605300
|
Spectrum Chemical Mfg Cor
PO11812KG |
12 kg | Poly Pail |
N/A
|
|
||||
18605301
|
Spectrum Chemical Mfg Cor
PO11845KGBL |
45 kg | Poly Drum |
N/A
|
|
||||
18605302
|
Spectrum Chemical Mfg Cor
PO118500LBBL |
500 lb. | Drum |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Polyethylene Glycol 1000, NF acts as a lubricant, coating the surfaces in aqueous and non-aqueous environments. All Spectrum Chemical NF products, including all NF polyethylene glycol products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
25322-68-3 | |
62.07 | |
LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
[H]OCCO |
(C2H4O)n | |
MFCD01779601 | |
ethane-1,2-diol |
Specifications
4.5 to 7.5 | |
1 | |
MFCD01779601 | |
LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
ethane-1,2-diol | |
NF | |
Poly Bottle |
25322-68-3 | |
(C2H4O)n | |
2.5 kg | |
[H]OCCO | |
62.07 | |
0.001 | |
16.0-19.0 cSt |