Learn More
Polyethylene Glycol 8000 (PEG), Fisher BioReagents™
Polyethylene Glycol 8000, HO-[CH2CH2O]n-H, CAS Number-25322-68-3, Polyethylene Oxide, Carbowax, PEG 8000, 1kg, Gray, White, 7000 to 9000, CAUTION!
$67.88 - $148.48
Chemical Identifiers
CAS | 25322-68-3 |
---|---|
Molecular Formula | (C2H4O)n |
Molecular Weight (g/mol) | 62.07 |
MDL Number | MFCD01779601 |
InChI Key | LYCAIKOWRPUZTN-UHFFFAOYSA-N |
Synonym | PEG 8000, Polyethylene Oxide, Carbowax™ |
IUPAC Name | ethane-1,2-diol |
SMILES | [H]OCCO |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
BP233-100
|
Fisher BioReagents
BP233100 |
100 g | Glass Bottle |
Each for $67.88
Case of 6 Each for $407.28
|
|
||||
BP233-1
|
Fisher BioReagents
FLBP2331 |
1 kg | Poly Bottle |
Each for $148.48
Case of 4 Each for $593.92
|
|
||||
Description
Polyethylene Glycol 8000, HO-[CH2CH2O]n-H, CAS Number-25322-68-3, Polyethylene Oxide, Carbowax, PEG 8000, 1kg, Gray, White, 7000 to 9000, CAUTION!
PEG 8000 is suitable as a medium for the fusion of mammalian cells. It is also used to precipitate bacteriophage from lysed cell supernatants.Chemical Identifiers
25322-68-3 | |
62.07 | |
LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
ethane-1,2-diol |
(C2H4O)n | |
MFCD01779601 | |
PEG 8000, Polyethylene Oxide, Carbowax™ | |
[H]OCCO |
Specifications
White | |
Solid | |
55°C | |
H(OCH2CH2)nOH | |
15, 7688 | |
100 g | |
[H]OCCO | |
62.07 | |
Pass Test | |
Polyethylene Glycol 8000 |
5.0 to 7.0 | |
25322-68-3 | |
(C2H4O)n | |
MFCD01779601 | |
PEG 8000, Polyethylene Oxide, Carbowax™ | |
LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
ethane-1,2-diol | |
7000 to 9000 | |
Glass Bottle |
Safety and Handling
CAUTION!
Emergency Overview
May cause skin, eye, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .
NFPA
Health:1
Flammability:1
Instability:0
Recommended Storage : RT