Learn More
Polyethyleneimine, branched, M.W. 10,000, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04033136
Description
Polyethyleneimine, branched, M.W. 10,000 acts as an auxiliary agent. It is used in paper-making, water treatment, detergents, adhesives and cosmetics. It is investigated as non-viral vector carriers in research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Yellow | |
M.W. 10,000 | |
9002-98-6 | |
MFCD00084427 | |
Soluble in toluene,methanol,ethanol,chloroform and xylene. Insoluble in water,alcohol,benzene,ethyl ether and acetone. | |
NOWKCMXCCJGMRR-UHFFFAOYSA-N | |
aziridine | |
9033 | |
99% |
Polyethyleneimine, branched | |
Liquid | |
((C2H5N)zC2H4N)y(C2H5N)x | |
ethyleneimine, ethylenimine, azacyclopropane, dimethyleneimine, ethylene imine, polyethyleneimine, dihydroazirene, everamine, aziran, polymin | |
500 g | |
*-CCNCCN(-*)CCN-* | |
43.07 | |
CHEBI:30969 | |
Amine-like |
Chemical Identifiers
9002-98-6 | |
43.07 | |
NOWKCMXCCJGMRR-UHFFFAOYSA-N | |
9033 | |
aziridine |
((C2H5N)zC2H4N)y(C2H5N)x | |
MFCD00084427 | |
ethyleneimine, ethylenimine, azacyclopropane, dimethyleneimine, ethylene imine, polyethyleneimine, dihydroazirene, everamine, aziran, polymin | |
CHEBI:30969 | |
*-CCNCCN(-*)CCN-* |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
DOTInformation : Hazard Class: 9; Packaging Group: III
RTECSNumber : TQ7915000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only