missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Polyethyleneimine, branched, M.W. 10,000, 99%, Thermo Scientific Chemicals

$92.96 - $1340.41

Chemical Identifiers

CAS 9002-98-6
Molecular Formula ((C2H5N)zC2H4N)y(C2H5N)x
Molecular Weight (g/mol) 43.07
MDL Number MFCD00084427
InChI Key NOWKCMXCCJGMRR-UHFFFAOYSA-N
Synonym ethyleneimine, ethylenimine, azacyclopropane, dimethyleneimine, ethylene imine, polyethyleneimine, dihydroazirene, everamine, aziran, polymin
PubChem CID 9033
ChEBI CHEBI:30969
IUPAC Name aziridine
SMILES *-CCNCCN(-*)CCN-*
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4033114
View Documents
Thermo Scientific Chemicals
04033114
25 g
Each for $92.96
 
AA4033122
View Documents
Thermo Scientific Chemicals
04033122
100 g
Each for $383.49
 
AA4033136
View Documents
Thermo Scientific Chemicals
04033136
500 g
Each for $1,340.41
 
Description

Description

Polyethyleneimine, branched, M.W. 10,000 acts as an auxiliary agent. It is used in paper-making, water treatment, detergents, adhesives and cosmetics. It is investigated as non-viral vector carriers in research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

9002-98-6
43.07
NOWKCMXCCJGMRR-UHFFFAOYSA-N
9033
aziridine
((C2H5N)zC2H4N)y(C2H5N)x
MFCD00084427
ethyleneimine, ethylenimine, azacyclopropane, dimethyleneimine, ethylene imine, polyethyleneimine, dihydroazirene, everamine, aziran, polymin
CHEBI:30969
*-CCNCCN(-*)CCN-*

Specifications

Yellow
9002-98-6
MFCD00084427
Soluble in toluene,methanol,ethanol,chloroform and xylene. Insoluble in water,alcohol,benzene,ethyl ether and acetone.
NOWKCMXCCJGMRR-UHFFFAOYSA-N
aziridine
9033
99%
Polyethyleneimine, branched, M.W. 10,000
Liquid
((C2H5N)zC2H4N)y(C2H5N)x
ethyleneimine, ethylenimine, azacyclopropane, dimethyleneimine, ethylene imine, polyethyleneimine, dihydroazirene, everamine, aziran, polymin
25 g
*-CCNCCN(-*)CCN-*
43.07
CHEBI:30969
Amine-like
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Hazard Class: 9; Packaging Group: III

RTECSNumber : TQ7915000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only