missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Justrite™ Polypropylene, Carboy with Sanitary Neck
For Use With: For storing and dispensing solvents, buffers, reagents and powders.; Autoclavable; Color: White (Translucent)
Supplier: Justrite™ 12935
Description
- Interchangeable caps and adapters provide top connection flexibility
- Rectangular ergonomic design
- Constructed from autoclavable USP Class VI, FDA grade materials
Specifications
±5% | |
2.6 gal. | |
White (Translucent) | |
For storing and dispensing solvents, buffers, reagents and powders. | |
10.75 in. | |
Threadless | |
Rectangular |
Autoclavable | |
10 L | |
273 x 163 x 488 mm | |
19.2 in. | |
Polypropylene | |
Carboy with Sanitary Neck | |
6.4 in. |