missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ Polypropylene, Carboy with Sanitary Neck

For Use With: For storing and dispensing solvents, buffers, reagents and powders.; Autoclavable; Color: White (Translucent)

$743.49 - $993.61

Specifications

Accuracy ±5%
Autoclavable Autoclavable
Color White (Translucent)
For Use With (Application) For storing and dispensing solvents, buffers, reagents and powders.
Material Polypropylene
View More Specs

Products 3
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
19-830-436
View Documents
Justrite™
12935
10 L
Each for $743.49
Only null left
 
19-830-437
View Documents
Justrite™
12936
20 L
Each for $781.41
Only null left
 
19-830-438
View Documents
Justrite™
12937
40 L
Each for $993.61
Only null left
 
Description

Description

  • Interchangeable caps and adapters provide top connection flexibility
  • Rectangular ergonomic design
  • Constructed from autoclavable USP Class VI, FDA grade materials
Specifications

Specifications

±5%
White (Translucent)
Polypropylene
Carboy with Sanitary Neck
Autoclavable
For storing and dispensing solvents, buffers, reagents and powders.
Threadless
Rectangular
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'