AirClean™ Systems Polypropylene Horizontal Laminar Flow Clean Bench
For Class 100 (ISO 5) applications, choose an Airclean Polypropylene Horizontal Laminar Flow Clean Bench.
Supplier: AirClean™ Systems AC4000HLF
Description
- UVTech™ microprocessor controller constantly monitors filter conditions
- Polypropylene construction
- Seamless, smooth construction minimizes air turbulence
- Built-in fluorescent lighting
- All-white surfaces are easy to clean
- Available in 110/220V versions
- Supplied fully assembled
Options
- Custom sturdy carts or stands
- Cup sink and vacuum fittings
- Polypropylene base cabinet
- Ultraviolet light source
- IV bar
This product will not protect the operator from biological hazards. Do not use this product with biological hazardous material.
- IV admixture preparation
- Drug compounding
- Plant cell culture
- Tissue culture (non-biological)
- Media preparation
- Pharmaceutical procedures
- Electronic assembly
Certifications
UL listed, CSA Certified
Compliance
Meets ISO 5 and FS209E Class 100 clean bench standards.
Specifications
41 in. | |
104.10 cm | |
48 in. |
27 in. | |
68.58 cm |