missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
AirClean™ Systems Polypropylene Horizontal Laminar Flow Clean Bench
For Class 100 (ISO 5) applications, choose an Airclean Polypropylene Horizontal Laminar Flow Clean Bench.
$17583.46 - $26258.84
Catalog Number | Mfr. No. | Height (English) | Height (Metric) | Width (English) | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Height (English) | Height (Metric) | Width (English) | Price | Quantity | |||
36-100-4382
|
AirClean™ Systems
AC5000HLF |
37 in. | 93.98 cm | 60 in. |
N/A
|
N/A | |||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
361004381
|
AirClean™ Systems
AC4000HLF |
41 in. | 104.10 cm | 48 in. |
N/A
|
N/A | |||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
361004384
|
AirClean™ Systems
AC8000HLF |
37 in. | 93.98 cm | 96 in. |
N/A
|
N/A | |||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
361004383
|
AirClean™ Systems
AC6000HLF |
37 in. | 93.98 cm | 72 in. |
N/A
|
N/A | |||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
- UVTech™ microprocessor controller constantly monitors filter conditions
- Polypropylene construction
- Seamless, smooth construction minimizes air turbulence
- Built-in fluorescent lighting
- All-white surfaces are easy to clean
- Available in 110/220V versions
- Supplied fully assembled
Options
- Custom sturdy carts or stands
- Cup sink and vacuum fittings
- Polypropylene base cabinet
- Ultraviolet light source
- IV bar
This product will not protect the operator from biological hazards. Do not use this product with biological hazardous material.
- IV admixture preparation
- Drug compounding
- Plant cell culture
- Tissue culture (non-biological)
- Media preparation
- Pharmaceutical procedures
- Electronic assembly
Certifications
UL listed, CSA Certified
Compliance
Meets ISO 5 and FS209E Class 100 clean bench standards.
Specifications
27 in. | |
68.58 cm |