missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Greiner Bio-One Polystyrene Vented Petri Dishes
Used for easy stacking. Greiner Bio-One Polystyrene Vented Petri Dishes is designed for improved gas exchange.
Supplier: Greiner Bio-One 628102
Description
- Vented for improved gas exchange
- Manufactured from crystal polystyrene for optimal analysis by microscopy
Specifications
Clear | |
Polystyrene | |
Non-sterile | |
600 | |
60 x 15 mm | |
60 mm |
Single-use | |
Round | |
Petri Dish | |
Yes | |
15 mm |
For Research Use Only. Not for use in diagnostic procedures, unless otherwise indicated.