missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Greiner Bio-One Polystyrene Vented Petri Dishes
Used for easy stacking. Greiner Bio-One Polystyrene Vented Petri Dishes is designed for improved gas exchange.
Supplier: Greiner Bio-One 633181
Description
- Vented for improved gas exchange
- Manufactured from crystal polystyrene for optimal analysis by microscopy
Specifications
Clear | |
Round | |
Petri Dish | |
Yes | |
16 mm |
Polystyrene | |
Sterile | |
480 | |
94 x 16 mm | |
94 mm |
For Research Use Only. Not for use in diagnostic procedures, unless otherwise indicated.