Polytetrafluoroethylene, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1261318
Description
PTFE may be used as a non stick coating. It can be used as engineering plastics. It is employed as lining reaction vessels, gaskets and pump packing, electrical insulator for high frequency applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Polytetrafluoroethylene | |
(C2F4)n | |
14,7562 | |
50 g | |
NaN |
9002-84-0 | |
MFCD00084455 | |
Insoluble in water. | |
FC(F)(-*)C(F)(F)-* |
Chemical Identifiers
9002-84-0 | |
NaN | |
FC(F)(-*)C(F)(F)-* |
(C2F4)n | |
MFCD00084455 |
Safety and Handling
RTECSNumber : KX4025000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only