missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Polytetrafluoroethylene, Thermo Scientific Chemicals

$109.58 - $1460.95

Chemical Identifiers

CAS 9002-84-0
Molecular Formula (C2F4)n
Molecular Weight (g/mol) NaN
MDL Number MFCD00084455
SMILES FC(F)(-*)C(F)(F)-*
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1261318
View Documents
Thermo Scientific Chemicals
A1261318
50 g
Each for $109.58
 
AAA1261330
View Documents
Thermo Scientific Chemicals
A1261330
250 g
Each for $365.23
 
AAA126130B
View Documents
Thermo Scientific Chemicals
A126130B
1000 g
Each for $1,460.95
 
Description

Description

PTFE may be used as a non stick coating. It can be used as engineering plastics. It is employed as lining reaction vessels, gaskets and pump packing, electrical insulator for high frequency applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

9002-84-0
NaN
FC(F)(-*)C(F)(F)-*
(C2F4)n
MFCD00084455

Specifications

9002-84-0
MFCD00084455
Insoluble in water.
FC(F)(-*)C(F)(F)-*
Polytetrafluoroethylene
(C2F4)n
14,7562
50 g
NaN
Videos
Safety and Handling

Safety and Handling

RTECSNumber : KX4025000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only