Polytetrafluoroethylene, Thermo Scientific Chemicals
Supplier:  Thermo Scientific Chemicals A1261330
 View more versions of this product
Catalog No. AAA1261330
Explore available promotions
Description
Description
PTFE may be used as a non stick coating. It can be used as engineering plastics. It is employed as lining reaction vessels, gaskets and pump packing, electrical insulator for high frequency applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Polytetrafluoroethylene
(C2F4)n
14,7562
250 g
NaN
9002-84-0
MFCD00084455
Insoluble in water.
FC(F)(-*)C(F)(F)-*
Chemical Identifiers
9002-84-0
NaN
FC(F)(-*)C(F)(F)-*
(C2F4)n
MFCD00084455
Videos
Safety and Handling
Safety and Handling
RTECSNumber : KX4025000
TSCA : Yes
Recommended Storage : Ambient temperatures