Learn More
Polyvinyl alcohol, 86-89% hydrolyzed, medium molecular weight, Thermo Scientific Chemicals
$68.85 - $176.60
Chemical Identifiers
CAS | 9002-89-5 |
---|---|
Molecular Formula | (C2H4O)n |
Molecular Weight (g/mol) | 44.05 |
MDL Number | MFCD00081922 |
InChI Key | IMROMDMJAWUWLK-UHFFFAOYSA-N |
Synonym | vinyl alcohol, polyvinyl alcohol, hydroxyethene, hydroxyethylene, ethenol, homopolymer, gohsenol, polyviol, elvanol, mowiol, poval |
PubChem CID | 11199 |
IUPAC Name | ethenol |
SMILES | OC(-*)C-* |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AA4123914
|
Thermo Scientific Chemicals
04123914 |
25 g |
Each for $68.85
|
|
||||||
AA4123922
|
Thermo Scientific Chemicals
04123922 |
100 g |
Each for $75.04
|
|
||||||
AA4123936
|
Thermo Scientific Chemicals
04123936 |
500 g |
Each for $176.60
|
|
||||||
Description
Polyvinyl alcohol is used as a biological reagent to test the dispersion of zinc oxide nanoparticles. It is an effective protein resistant material and is used to stabilize gold colloids of diameter 3-8nm. It is used as an emulsion polymerization aid to make polyvinyl acetate, as a protective colloid, as a film used in the water transfer printing process, as a surfactant for the formation of polymer encapsulated nanobeads, as an embolization agent in medical procedures and as a fixative for specimen collection.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
9002-89-5 | |
44.05 | |
IMROMDMJAWUWLK-UHFFFAOYSA-N | |
11199 | |
OC(-*)C-* |
(C2H4O)n | |
MFCD00081922 | |
vinyl alcohol, polyvinyl alcohol, hydroxyethene, hydroxyethylene, ethenol, homopolymer, gohsenol, polyviol, elvanol, mowiol, poval | |
ethenol |
Specifications
Powder | |
(C2H4O)n | |
vinyl alcohol, polyvinyl alcohol, hydroxyethene, hydroxyethylene, ethenol, homopolymer, gohsenol, polyviol, elvanol, mowiol, poval | |
25 g | |
OC(-*)C-* | |
44.05 | |
86 to 89% | |
Polyvinyl alcohol, Medium molecular weight, Hydrolyzed |
9002-89-5 | |
MFCD00081922 | |
Soluble in water. | |
IMROMDMJAWUWLK-UHFFFAOYSA-N | |
ethenol | |
11199 | |
Odorless |
Safety and Handling
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only