Learn More
Potassium bisulfate, 98.5%, for analysis
CAS: 7646-93-7 | HKO4S | 136.16 g/mol
$265.21 - $265.21
Identifiants chimiques
| CAS | 7646-93-7 |
|---|---|
| Molecular Formula | HKO4S |
| Molecular Weight (g/mol) | 136.16 |
| MDL Number | MFCD00011404 |
| InChI Key | CHKVPAROMQMJNQ-UHFFFAOYSA-M |
| Synonym | potassium bisulfate, potassium hydrogen sulfate, potassium bisulphate, sulfuric acid, monopotassium salt, potassium acid sulfate, sal enixum, potassium hydrogensulphate, monopotassium sulfate, acid potassium sulfate, caswell no. 682c |
| PubChem CID | 516920 |
| IUPAC Name | potassium hydrogen sulfate |
| SMILES | [K+].OS([O-])(=O)=O |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | ||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | ||||
|
AC207755000
|
Thermo Scientific Chemicals
207755000 |
500 g | Glass bottle |
chaque for $265.21
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identifiants chimiques
| 7646-93-7 | |
| 136.16 | |
| CHKVPAROMQMJNQ-UHFFFAOYSA-M | |
| 516920 | |
| [K+].OS([O-])(=O)=O |
| HKO4S | |
| MFCD00011404 | |
| potassium bisulfate, potassium hydrogen sulfate, potassium bisulphate, sulfuric acid, monopotassium salt, potassium acid sulfate, sal enixum, potassium hydrogensulphate, monopotassium sulfate, acid potassium sulfate, caswell no. 682c | |
| potassium hydrogen sulfate |
Spécifications
| 214.0°C | |
| White | |
| 500 g | |
| HKO4S | |
| MFCD00011404 | |
| 15, 7734 | |
| Solubility in water: 490g/L (20°C). Other solubilities: decomposes in alcohol | |
| [K+].OS([O-])(=O)=O | |
| 136.16 | |
| 136.16 | |
| Analytical | |
| 2.32 |
| 7646-93-7 | |
| Adhering Crystals | |
| 98.50% | |
| KHSO4 | |
| 01,909 | |
| potassium bisulfate, potassium hydrogen sulfate, potassium bisulphate, sulfuric acid, monopotassium salt, potassium acid sulfate, sal enixum, potassium hydrogensulphate, monopotassium sulfate, acid potassium sulfate, caswell no. 682c | |
| CHKVPAROMQMJNQ-UHFFFAOYSA-M | |
| potassium hydrogen sulfate | |
| 516920 | |
| 98.5% | |
| Glass bottle | |
| Potassium bisulfate, c.p. |
Sécurité et manipulation
GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 231-594-1
missing translation for 'rtecsNumber' : TS7200000
missing translation for 'tsca' : TSCA
RUO – Research Use Only